Learn More
Kd Medical Inc SUPER BROTH+PHOSPHATES BOTTLE
SDP
Catalog No. 501018957
Encompass_Preferred
Super Optimal Broth (SOB medium) is a nutrient-rich bacterial growth medium used for microbiological culture, generally of Escherichia coli. One 500ml bottle.
Catalog No. 50-101-8957 Supplier Kd Medical Inc Supplier No. BLE3160
Super Optimal Broth (SOB medium) is a nutrient-rich bacterial growth medium used for microbiological culture, generally of Escherichia coli. One 500ml bottle.