Learn More
Teknova Super Broth with Animal-Free Soytone. 1000mL, Sterile.
SDP
Catalog No. 502387691
Encompass
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.
Catalog No. 50-238-7691 Supplier Teknova Supplier No. S1530
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.