Learn More
Leica Biosystems Leica Super Cassettes, Orange - 3 x 2 x 3/4 in (75 x 52 x 17 mm), 100/Case
Catalog No. NC9886464
Encompass_Preferred
Leica Biosystems Super Cassettes (orange) are oversized histology processing and embedding cassettes designed to accommodate larger tissue specimens and streamline laboratory workflows.
  • Allow processing and embedding of larger specimens.
  • Color coded for easy sample identification.
  • Packaged 100 per case for inventory efficiency.
  • Compatible with standard histology processing equipment.
  • Dimensions: 3 x 2 x 3/4 in (75 x 52 x 17 mm).
Catalog No. NC9886464 Supplier Leica Biosystems Supplier No. 38VSP59061
Leica Biosystems Super Cassettes (orange) are oversized histology processing and embedding cassettes designed to accommodate larger tissue specimens and streamline laboratory workflows.
  • Allow processing and embedding of larger specimens.
  • Color coded for easy sample identification.
  • Packaged 100 per case for inventory efficiency.
  • Compatible with standard histology processing equipment.
  • Dimensions: 3 x 2 x 3/4 in (75 x 52 x 17 mm).