Learn More
Leica Biosystems Leica Biosystems White Super Cassettes, 100/Case - Histology tissue cassettes
Catalog No. NC0920346
Encompass_Preferred
Leica Biosystems White Super Cassettes are disposable tissue cassettes intended for routine histology processing, embedding, and short-term specimen storage. Packaged 100 per case to support consistent specimen containment and efficient laboratory workflows.
  • White color for easy specimen visibility.
  • Includes 100 cassettes per case.
  • Compatible with standard tissue processors and embedding stations.
  • Dimensions: 3 x 2 x 3/4 in.
  • Designed for routine biopsy and surgical specimens.
Catalog No. NC0920346 Supplier Leica Biosystems Supplier No. 38VSP59060
Leica Biosystems White Super Cassettes are disposable tissue cassettes intended for routine histology processing, embedding, and short-term specimen storage. Packaged 100 per case to support consistent specimen containment and efficient laboratory workflows.
  • White color for easy specimen visibility.
  • Includes 100 cassettes per case.
  • Compatible with standard tissue processors and embedding stations.
  • Dimensions: 3 x 2 x 3/4 in.
  • Designed for routine biopsy and surgical specimens.