Learn More
MilliporeSigma™ Supelco™ Super Clean™ (Base-Plate Design) Gas Purifiers
Shop All MilliporeSigma Supelco ProductsDescription
- Provides gas at the proper purity level and pressure, based on its intended use
- The purifiers connect into a base plate in such a manner that gas supply is not interrupted during purifier change-out
- Purifier can be replaced without shutting down the entire system
- Eliminates the risk of forcing a slug of air into the instrument after purifier change-out
Specifications
Specifications
| For Use With (Application) | Enhancing The Performance of GC Detectors |
| For Use With (Equipment) | GC Detectors |
| Quantity | 1 Ea. |
| Description | For Fuel Gas |
| Type | Dual Trap (Hydrocarbon, Moisture) |