Learn More
MilliporeSigma™ Supelco™ Super Clean™ (Base-Plate Design) Kits
Catalog No. 111008374 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Product Type:
Carrier Gas Kit
Helium Carrier Gas Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
11-100-8374 Carrier Gas Kit
11-100-8040 Helium Carrier Gas Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-8374 Supplier MilliporeSigma™ Supelco™ Supplier No. 28878U
Add to Cart
Add to Cart
A unique, point-of-use diffusion proof purification system to purify carrier, fuel, and other gases for GC or GC-MS systems
  • Purifiers remove hydrocarbons, oxygen (color indicated), and moisture (color indicated) to better than 6.0 gas (99.9999%) quality at 2 L/min., independent of the original gas quality
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Purification of Carrier, Fuel, and Other Gases
For Use With (Equipment) GC or GC-MS Systems
Includes SU861026, and SU861011
Product Type Carrier Gas Kit
Super Clean is a trademark of Scientific Glass Technology B.V.