Learn More
Med Vet International Super Cut Metzenbaum Scissors, Curved. 5 3/4" (14cm), J0911CA, Each
Catalog No. 5000584203
Encompass_Preferred
Super Cut Scissors. These scissors have a specially designed razor sharp edge that cuts through tissue like butter. The lower blade has micro fine serrations to prevent tissue slippage. Black finger rings for easy identification. Metzenbaum Scissors. Curved. 5 3/4" (14cm).
Catalog No. 50-005-84203 Supplier Med Vet International Supplier No. JOR911CA
Super Cut Scissors. These scissors have a specially designed razor sharp edge that cuts through tissue like butter. The lower blade has micro fine serrations to prevent tissue slippage. Black finger rings for easy identification. Metzenbaum Scissors. Curved. 5 3/4" (14cm).