Learn More
NEOGENESTAR LLC Super Deep 24 Well Plate For 25ml, Polypropylene, 24 Plates/CS
Catalog No. NC1831972
Encompass_Preferred
Nonsterile, autoclavable (with sandwich cover on top, if desired) Volume: 25mL, recommended culture volume: 5-7 ml, Polypropylene 3 plates / bag, 24 plates / CS
Catalog No. NC1831972 Supplier NEOGENESTAR LLC Supplier No. NGS 24 WELL 25 ML
Nonsterile, autoclavable (with sandwich cover on top, if desired) Volume: 25mL, recommended culture volume: 5-7 ml, Polypropylene 3 plates / bag, 24 plates / CS