Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Super-DHB suitable for matrix substance for MALDI-MS, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014995 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
10 x 10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4995 10 x 10 mg
11-101-4993 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4995 Supplier MilliporeSigma Supelco Supplier No. 5086210X10MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The “super-DHB” matrix consists of a 9:1 (w/w) mixture of 2,5-DHB and 2-hydroxy-5-methoxybenzoic acid, respectively, since this ratio was found to be the most effective. The beneficial effect of the 2-hydroxy- 5-methoxybenzoic acid additive is attributed to a disorder in the 2,5-DHB crystal lattice resulting in enhanced ion desorption (i.e. ‘softer’ desorption) and reduced internal energy, which in turn results in reduced metastable fragmentation. An increase in sensitivity of up to 2 to 3-fold has been reported for a standard dextran mixture with concomitant enhanced resolution attributed to reduced metastable ion formation. The super-DHB matrix has also been reported to generate enhanced signals from tryptic glycopeptides. The use of super-DHB matrix is reported to reduce the presence of matrix background ions and result in glycan signals exhibiting better signal to noise ratios and higher resolution in the mass spectrum.
TRUSTED_SUSTAINABILITY

Specifications

CAS 63542-76-7
Purity Grade Notes 9:1 mixture of DHB and 2-hydroxy-5-methoxybenzoic acid
Solubility Soluble in methanol
Type Organic Standard
Quantity 10 x 10 mg
Percent Purity ≥99% (sum of DHB and 2-hydroxy-5-methoxybenzoi c acid, HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.