Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro Super Erecta 27UPS SiteSelect Mobile-Ready Industrial Wire Shelving Post

Catalog No. 13450001 Shop All Metro Products
Change view
Click to view available options
Diameter (Metric):
1 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
13-450-001 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-450-001 Supplier Metro Supplier No. 27UPS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metro Super Erecta SiteSelect Posts provide a visual guide for postioning and adjusting shelves.

  • An easy-to-identify double groove every 8 in. offers a quick way to align all shelves.
  • Stationary posts include an adjustable leveling foot.
  • Mobile posts come without leveling bolt assembly to accommodate stem casters.
  • Type 304 Polished Stainless Steel finish addresses the most aggressive applications and environments.

Specifications

Product Type Mobile Shelving Post
Color Silver
Material Stainless Steel
Diameter (Metric) 1 in.
For Use With (Application) Mobile Post for Shelving Unit
For Use With (Equipment) Metro Super Erecta Shelving
Includes Mobile-Ready Post
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.