Learn More
Metro Super Erecta Mobile Security Shelving Unit
Catalog No. 01001055 Shop All Metro Products
Change view
Click to view available options
Dimensions (L x W x H):
28.5 x 52.75 x 67.9375 in.
28.5 x 65 x 68.4375 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
01-001-055 28.5 x 65 x 68.4375 in.
01-001-035 28.5 x 52.75 x 67.9375 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-001-055 Supplier Metro Supplier No. SEC56VK3SL
Add to Cart
Add to Cart
Metro Super Erecta Mobile Security Shelving Unit with SlideSecure Sliding Door.
  • Keep sensitive items safe from loss or pilferage.
  • Heavy-gauge open wire construction keeps contents visible at all times, making it easy to check inventory.
  • Metro SlideSecure Space Saving Doors provide the same level of protection as traditional security units with the space saving convenience of our sliding door system.
  • Units include two chrome Super Erecta shelves which must be used as the top and bottom shelves of the unit.
  • Add optional intermediate shelves for increased storage capabilities.
  • Includes four 5 in. diameter casters with plated finish and polyurethane tread (two swivel and two swivel with brakes).
  • Padlock sold separately.
Specifications
Product Type Mobile Shelving Post
Color Gray
Material Epoxy-coated Steel
Autoclavable Not Autoclavable
Dimensions (L x W x H) 28.5 x 65 x 68.4375 in.
With Casters Yes