Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro Super Erecta Solid Shelf

Catalog No. 01001469 Shop All Metro Products
Change view
Click to view available options
Dimensions (L x W):
18 x 24 in.
18 x 30 in.
18 x 36 in.
18 x 42 in.
18 x 48 in.
18 x 60 in.
24 x 24 in.
24 x 30 in.
24 x 36 in.
24 x 42 in.
24 x 48 in.
24 x 60 in.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
01-001-469 24 x 60 in.
01-001-354 18 x 24 in.
01-001-357 18 x 30 in.
01-001-362 18 x 36 in.
01-001-366 18 x 42 in.
01-001-375 18 x 48 in.
01-001-379 18 x 60 in.
01-001-382 24 x 24 in.
01-001-401 24 x 30 in.
01-001-407 24 x 36 in.
01-001-448 24 x 42 in.
01-001-453 24 x 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 01-001-469 Supplier Metro Supplier No. 2460NFST316
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Super Erecta Solid Shelf are made of Premium Autoclavable/Cart-Washable Type 316 Stainless Steel.

  • Engineered for the harshest environments, these premium shelves are constructed entirely from type 316 stainless steel, offering unparalleled resistance to corrosion, chemicals, and high temperatures.
  • Ideal for commercial labs and industrial applications, these shelves provide exceptional durability and reliability in the most demanding conditions.
  • Built to withstand the toughest environments, these fully type 316 stainless steel shelves deliver long-lasting performance and protection.

Specifications

Product Type Extra Shelf
CAS 67-56-1, 1189505-24-5
Autoclavable Autoclavable
Dimensions (L x W) 24 x 60 in.
Linear Formula C17H8D8ClN3O2
MDL Number MFCD28898900
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
No. of Shelves 1
Formula Weight 337.83
With Casters No
Packaging Ampule
Quantity 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.