Learn More
Metro Super Erecta Stationary Security Shelving Unit
Catalog No. 01001883 Shop All Metro Products
Change view
Click to view available options
Dimensions (L x W x H):
28.5 x 50.5 x 66.8125 in.
28.5 x 62.5 x 66.8125 in.
Material:
Chrome
Epoxy-coated Steel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Material
01-001-883 28.5 x 50.5 x 66.8125 in. Chrome
01-001-041 28.5 x 62.5 x 66.8125 in. Chrome
01-001-046 28.5 x 62.5 x 66.8125 in. Epoxy-coated Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-001-883 Supplier Metro Supplier No. SEC55CSL
Add to Cart
Add to Cart
Metro Super Erecta Stationary Security Shelving Unit with SlideSecure Sliding Door.
  • Keep sensitive items safe from loss or pilferage.
  • Heavy-gauge open wire construction keeps contents visible at all times, making it easy to check inventory.
  • Metro SlideSecure Space Saving Doors provide the same level of protection as traditional security units with the space saving convenience of our sliding door system.
  • Units include two Super Erecta shelves which must be used as the top and bottom shelves of the unit.
  • Add optional intermediate shelves for increased storage capabilities.
  • Padlock sold separately.
Specifications
Product Type Rack
Color Gray
Material Chrome
Autoclavable Not Autoclavable
Dimensions (L x W x H) 28.5 x 50.5 x 66.8125 in.
With Casters No