Learn More
Waters Corp Waters Ferrule, Super Flangeless, Stainless Steel Ring, 5 per pack
Catalog No. 50466656 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement ferrule, super flangeless, designed with a stainless steel ring for robust performance. It is commonly used in Mass Spectrometer Fluidics applications.
  • Super flangeless design.
  • Constructed with a stainless steel ring.
  • Supplied in a pack of 5.
  • Suitable for mass spectrometer fluidics.
  • Functions as a fitting.
  • Designed for MS system types.
Catalog No. 50-466-656 Supplier Waters Corp Supplier No. 700005489
This product is a replacement ferrule, super flangeless, designed with a stainless steel ring for robust performance. It is commonly used in Mass Spectrometer Fluidics applications.
  • Super flangeless design.
  • Constructed with a stainless steel ring.
  • Supplied in a pack of 5.
  • Suitable for mass spectrometer fluidics.
  • Functions as a fitting.
  • Designed for MS system types.