Learn More
Roboz Surgical Super Fine #5SF Dumonts
SDP
Catalog No. NC1532347
Encompass
These forceps have the finest tips ever produced by Dumont. They are half the size of the regular #5 biologie tips.
This item is not returnable. View return policy
Catalog No. NC1532347 Supplier Roboz Surgical Supplier No. RS4955
These forceps have the finest tips ever produced by Dumont. They are half the size of the regular #5 biologie tips.