Learn More
IDEX Health & Science Upchurch SUPER FLANGELESS FERRULE 1/16"
Catalog No. NC1792649
Encompass_Preferred
Super Flangeless™ Ferrule w/SST Ring, 1/4-28 Flat-Bottom, for 1/16" OD Natural - Single
Catalog No. NC1792649 Supplier IDEX Health & Science Upchurch Supplier No. P250
Super Flangeless™ Ferrule w/SST Ring, 1/4-28 Flat-Bottom, for 1/16" OD Natural - Single