Learn More
Analytical Sales & Services Super Flangeless M6 Nut for 1/16” Tubing
Catalog No. 5000804668
Encompass
Super Flangeless Fitting - M6 Black 1/16'' Flangeless Nut
¥ Rated to 2400psi on PEEKª tubing
Qty/Pkg: 10PK
Catalog No. 50-008-04668 Supplier Analytical Sales & Services Supplier No. 59207X
Super Flangeless Fitting - M6 Black 1/16'' Flangeless Nut
¥ Rated to 2400psi on PEEKª tubing
Qty/Pkg: 10PK