Learn More
Charles Supper Co Inc Super Glue Gel, 2g tubes, 2/pack
SDP
Catalog No. NC0464418
Encompass
Permanently seals our capillary tubes, mount glass fibers to brass specimen pins or any other use that requires a permanent bond.
Catalog No. NC0464418 Supplier Charles Supper Co Inc Supplier No. SG08
Permanently seals our capillary tubes, mount glass fibers to brass specimen pins or any other use that requires a permanent bond.