Learn More
International Enviroguard™ Super Heavy Duty 3417 CPE Shoe Covers
Catalog No. 19472491 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-472-491 White
19-472-492 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-472-491 Supplier International Enviroguard™ Supplier No. 3417
Add to Cart
Add to Cart
Water resistant shoe covers feature a strong elastic closure to help protect shoes from liquids and debris. Ideal for a variety of tough environments that require heavy duty protection.
CPE, disposable white shoe covers help protect shoes and surrounding environments from dust, dirt, grime, and liquids. They're more durable than polypropylene, so you're protected in tough environments. Whether you're in a cleanroom, a hospital, or working at a commercial site, these durable shoe covers help protect against chemicals, dirt, dust, liquid splash, non-hazardous particulates, and even shallow immersion.
  • Water-resistant and come with a strong elastic closure
  • One size fits most; fits up to a men's size 12 1/2 shoe
  • Can be worn in indoor and outdoor environments
  • Possible applications include healthcare, controlled environments, food processing, pharmaceuticals, construction, realtors, movers, contract work, and more
Specifications
Size X-Large
Material CPE
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Standard
Packaging Quantity 300/Cs.
Resistance Dry Particulates, Splash
Product Type Controlled Environments Shoe Covers