Learn More
Research Products International Corp Super HT PAP Pen, Mini, 2.5mm Tapered Tip offers over 400 Applications
SDP
Catalog No. 50550222
Encompass
The PAP Pen draws thin film-like, hydrophobic barriers on microscope slides. This barrier creates the proper surface tension to hold antibody solutions and other specimens within a target area. The PAP Pen releases fluid in precise amounts to provide optimum surface tension. This eliminates the possibility of flooding the slide.
The PAP Pen can be effectively used for immunostaining by the PAP (peroxidase-Antiperoxidase), ABC, frozen section and fluorescent antibodies methods. If the frozen section method is used, two specimens are easily separated by applying two individual circles on the same slide.
WARNING! This product can expose you to 1-Bromopropane, a chemical which is known to the State of California to cause developmental reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-550-222 Supplier Research Products International Corp Supplier No. 195506
The PAP Pen draws thin film-like, hydrophobic barriers on microscope slides. This barrier creates the proper surface tension to hold antibody solutions and other specimens within a target area. The PAP Pen releases fluid in precise amounts to provide optimum surface tension. This eliminates the possibility of flooding the slide.
The PAP Pen can be effectively used for immunostaining by the PAP (peroxidase-Antiperoxidase), ABC, frozen section and fluorescent antibodies methods. If the frozen section method is used, two specimens are easily separated by applying two individual circles on the same slide.
WARNING! This product can expose you to 1-Bromopropane, a chemical which is known to the State of California to cause developmental reproductive harm. For more information go to www.P65Warnings.ca.gov.