Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Super-Hydride™ Solution

Catalog No. 1797288L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
8 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-972-88L 8 L
17-972-8800ML 800 mL
50-308-1435 4 x 25 mL
17-972-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 17-972-88L Supplier Sigma Aldrich Supplier No. 1797288L
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Super-Hydride™ solution (Lithium triethylborohydride or LiTEBH) is widely used as a powerful and selective reducing agent that shows super hydride activity in organic synthesis.LiTEBH can be used as a reagent: To reduce alkyl halides to alkanes via dehydrogenation reactions.For the selective reduction of epoxides to Markovnikov alcohols. To reduce tosylates or mesylates primary alcohols to hydrocarbons.For reductive cyclization reactions for the preparation of useful intermediates.In the synthesis of hepta(manno-3-deoxy-6-O-t-butyldimethylsilyl)-+a-cyclodextrin by reduction of hepta(manno-2,3-anhydro-6-O-t-butyldimethylsilyl)-+a-cyclodextrin.For hydrodefluorination of C-F bonds using Ni catalyst.To prepare alkynyl alcohols from cleavage of cyclic keto-vinyl triflates.In the stereoselective reduction of bicyclic imides, isoquinolines, and pyridines.To prepare tungsten and molybdenum hydride complexes.Super-Hydride is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 22560-16-3
Density 0.892 g/mL at 25°C
Molecular Formula C6H16BLi
Linear Formula Li(C2H5)3BH
MDL Number MFCD00011703
Quantity 8 L
Synonym Lithium triethylborohydride solution
Molecular Weight (g/mol) 105.94 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.