Learn More
MSC Betco Degreaser, Heavy-Duty, Butyl-Based, Cherry Fragrance, 55 gal
Catalog No. 502819518
Encompass
Super Kemite Degreaser is a heavy-duty cleaner/degreaser designed for tough jobs. It is formulated to instantly dissolve and emulsify heavy grease, carbon deposits, rubber burns, oil, ink, and dirt accumulations. It can be sprayed or mopped on and then rinsed off.
  • Concentrated butyl-based cleaner for light to heavy-duty cleaning
  • Deodorizes with a cherry fragrance as it cleans
  • Versatile dilution, suitable for light or heavy-duty cleaning
  • Low foaming, ideal for automatic scrubbers and pressure washers
Catalog No. 50-281-9518 Supplier MSC Supplier No. 72656051
Super Kemite Degreaser is a heavy-duty cleaner/degreaser designed for tough jobs. It is formulated to instantly dissolve and emulsify heavy grease, carbon deposits, rubber burns, oil, ink, and dirt accumulations. It can be sprayed or mopped on and then rinsed off.
  • Concentrated butyl-based cleaner for light to heavy-duty cleaning
  • Deodorizes with a cherry fragrance as it cleans
  • Versatile dilution, suitable for light or heavy-duty cleaning
  • Low foaming, ideal for automatic scrubbers and pressure washers