Learn More
Ted Pella Inc HISTOBOND + Super Mega Slides, 75.2 x 50.4 x 1mm (3" x 2" x 1mm) 50/box
SDP
Catalog No. NC1811932
Encompass
HISTOBOND + Super Mega Slides have a thin white frosted end for labeling. The unfrosted area is sized to accommodate the standard large section cut from a paraffin embedded tissue block from the large Super Mega and Super Mega Slim Base Molds (60 x 45mm).
Catalog No. NC1811932 Supplier Ted Pella Inc Supplier No. 260240
HISTOBOND + Super Mega Slides have a thin white frosted end for labeling. The unfrosted area is sized to accommodate the standard large section cut from a paraffin embedded tissue block from the large Super Mega and Super Mega Slim Base Molds (60 x 45mm).