Learn More
Biogenex Laboratories SuperMount Permanent Aqueous Mounting Medium
SDP
Catalog No. NC9742697
Encompass_Preferred
SuperMount is compatible with most chromogens commonly used in immunohistochemistry and in situ hybridization including AEC, DAB, Fast Red, New Fuchsin, and BCIP/NBT. SuperMount is also compatible with specimens stained with fluorescent dyes, including rhodamine, fluorescein, Texas Red, and even with phycoerythrin and its related derivatives such as phycocyanin and allophycocyanin. SuperMount can also, be used for the mounting of special stain such as Fat Stain. Oil Red O. (Note: SuperMountis not recommended for use with Nuclear Fast Red, eosin, or Methyl Green, as color may fade during long-term storage). SuperMount can be used for the mounting of all biological specimens, including stained tissue sections, cytospin preparations, and blood smears. This mounting medium can be used in laboratory settings without a fume hood.
Catalog No. NC9742697 Supplier Biogenex Laboratories Supplier No. HK0797K
SuperMount is compatible with most chromogens commonly used in immunohistochemistry and in situ hybridization including AEC, DAB, Fast Red, New Fuchsin, and BCIP/NBT. SuperMount is also compatible with specimens stained with fluorescent dyes, including rhodamine, fluorescein, Texas Red, and even with phycoerythrin and its related derivatives such as phycocyanin and allophycocyanin. SuperMount can also, be used for the mounting of special stain such as Fat Stain. Oil Red O. (Note: SuperMountis not recommended for use with Nuclear Fast Red, eosin, or Methyl Green, as color may fade during long-term storage). SuperMount can be used for the mounting of all biological specimens, including stained tissue sections, cytospin preparations, and blood smears. This mounting medium can be used in laboratory settings without a fume hood.