Learn More
Electron Microscopy Sciences SUPER-NUOVA™ 6" Probe for Hot Plate, heating surface, RPM
SDP
Catalog No. 501899848
Encompass_Preferred
Super-Nuova hot plate stirer remote probe, 6" (15.2cm), 316 stainless steel. Has a maximum temperature of 1100ºC.
Catalog No. 50-189-9848 Supplier Electron Microscopy Sciences Supplier No. 7235201
Super-Nuova hot plate stirer remote probe, 6" (15.2cm), 316 stainless steel. Has a maximum temperature of 1100ºC.