Learn More
Ted Pella Inc Super PAP Pen Liquid Blocker, regular size, approximately 16.5ml, tip size 5mm
SDP
Catalog No. NC1710579
Encompass
PAP Pens are used for immunohistochemical applications. The hydrophobic properties allow the user to draw barriers on the slide, in any pattern, to confine the flow of reagents to a defined area. A minimum of valuable antiserum will be used. Visible marking is light green/blue and can be removed with Xylene. For immunohistochemistry, the PAP Pen should be applied after deparaffinization.
Catalog No. NC1710579 Supplier Ted Pella Inc Supplier No. 22309
Add to Cart
Edge
Add to Cart
PAP Pens are used for immunohistochemical applications. The hydrophobic properties allow the user to draw barriers on the slide, in any pattern, to confine the flow of reagents to a defined area. A minimum of valuable antiserum will be used. Visible marking is light green/blue and can be removed with Xylene. For immunohistochemistry, the PAP Pen should be applied after deparaffinization.