Learn More
Ihc World LLC Super Pap Pen Liquid Blocker For IHC, Small, Each
SDP
Catalog No. NC0552848
Encompass_Preferred
This ready-to-use hydrophobic barrier pens is designed for immunohistochemistry applications. The surface tension provided by the circle, square, or any other shapes drawn to fit tissue sections with the pen ensures that only the amount of antibody s
Catalog No. NC0552848 Supplier Ihc World LLC Supplier No. SPM0928
This ready-to-use hydrophobic barrier pens is designed for immunohistochemistry applications. The surface tension provided by the circle, square, or any other shapes drawn to fit tissue sections with the pen ensures that only the amount of antibody s