Learn More
Ihc World LLC SUPER PAT PEN LARGE
SDP
Catalog No. NC0200055
Encompass
This ready-to-use hydrophobic barrier pens is designed for immunohistochemistry applications. The surface tension provided by the circle, square, or any other shapes drawn to fit tissue sections with the pen ensures that only the amount of antibody s
Catalog No. NC0200055 Supplier Ihc World LLC Supplier No. SPR0905
This ready-to-use hydrophobic barrier pens is designed for immunohistochemistry applications. The surface tension provided by the circle, square, or any other shapes drawn to fit tissue sections with the pen ensures that only the amount of antibody s