Learn More
System Biosciences LLC Super PiggyBac Transposase Expression Vector
SDP
Supplier:  System Biosciences LLC PB210PA1
Encompass
The Super PiggyBac Transposase Expression Vector from System Biosciences is a plasmid vector that expresses the hyperactive PiggyBac transposase gene for efficient transposon-mediated gene delivery and stable integration in mammalian cells. This vector contains the CMV promoter to drive strong expression in mammalian cells. The PiggyBac transposase catalyzes the movement of a gene of interest flanked by PiggyBac inverted terminal repeats into the host genome for stable transgene integration and expression. This vector is useful for gene delivery, transgenesis, gene therapy, and genome engineering applications.
Catalog No. NC2261624