Learn More
MSC 3M Super Polish Pad 4100, White, Polyester Fiber
Catalog No. 502839731
Encompass
This super polish pad is designed for light cleaning, buffing, and polishing, effectively removing scuffs and soil without dulling floor finishes. Constructed from durable polyester fibers in an open-textured, non-woven form, it ensures consistent performance for floor maintenance.
  • Removes soil and scuff marks; buffs and polishes surfaces
  • Can be used for spray cleaning or dry buffing soft finishes
  • Made from durable polyester fibers for longevity
  • Washable and reusable, reducing replacement costs
  • Uniform coating prevents scratching and ensures consistent results
  • Suitable for low-speed floor machines and automatic scrubbers on various finish-coated floors
Catalog No. 50-283-9731 Supplier MSC Supplier No. 75619635
This super polish pad is designed for light cleaning, buffing, and polishing, effectively removing scuffs and soil without dulling floor finishes. Constructed from durable polyester fibers in an open-textured, non-woven form, it ensures consistent performance for floor maintenance.
  • Removes soil and scuff marks; buffs and polishes surfaces
  • Can be used for spray cleaning or dry buffing soft finishes
  • Made from durable polyester fibers for longevity
  • Washable and reusable, reducing replacement costs
  • Uniform coating prevents scratching and ensures consistent results
  • Suitable for low-speed floor machines and automatic scrubbers on various finish-coated floors