Learn More
Research Products International Corp Super Polyfoil Autoclaving Wrap, 4 x 4 Inch, 100 Sheets per Package
SDP
Catalog No. 501367932
Encompass
Super Polyfoil creates a chemically inert barrier to cover vessels for autoclaving or incubation and storage. Super Polyfoil features a thermoplastic backing laminated to 1mil thick foil. This superb combination allows material to be easily folded without puncturing or tearing. The polyester film also provides an inert barrier between the foil and liquids for better resistance to acids or caustic reagents. Complies with FDA regulations governing direct food contact applications.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-136-7932 Supplier Research Products International Corp Supplier No. 246415
Super Polyfoil creates a chemically inert barrier to cover vessels for autoclaving or incubation and storage. Super Polyfoil features a thermoplastic backing laminated to 1mil thick foil. This superb combination allows material to be easily folded without puncturing or tearing. The polyester film also provides an inert barrier between the foil and liquids for better resistance to acids or caustic reagents. Complies with FDA regulations governing direct food contact applications.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.