Learn More
Akro Mils Inc SUPER SIZE AKROBIN GREY MOQ4
Catalog No. NC2302823
Encompass_Preferred
Super Size AkroBin 23-7/8 x 8-1/4 x 7, Grey.Priced per each. Minimum order quantity 4. Order in multiples of 4.
Catalog No. NC2302823 Supplier Akro Mils Inc Supplier No. 30284GREY
Super Size AkroBin 23-7/8 x 8-1/4 x 7, Grey.Priced per each. Minimum order quantity 4. Order in multiples of 4.