Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Super Snowflake Stencils
SDPSCI_ED

Catalog No. S13219
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13219 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13219 Supplier Roylco™ Supplier No. R58622
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Let nature be your guide as children learn to appreciate the value of originality through these snowflake stencils.

Just like snowflakes, no two children are alike. Take advantage of a brisk winter's day by teaching your students about the value of uniqueness with these fun snowflake stencils.

  • Teaches children to observe the world around them
  • Notice how colors play an important role in distinguishing the seasons

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Format Kit
Includes Stencils, Idea Guide
Quantity 12 Stencils
Languages English, Spanish, French, German
Subject Earth Science - Weather and Climate
Diameter (English) 8 in.
Diameter (Metric) 20 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.