Learn More
WYK Sorbents LLC Safety Sorbent, 2LB bag, 12/case
SDP
Catalog No. NC0100450
Encompass_Preferred
Anti-Slip Safety Sorbent is concentrated and very economical to use. Studies have shown the average bag is used twelve times in cleaning up small spills and nuisance drips. A 510 bag will absorb up to 3 US gallons of oil and most other liquids.NON BIODEGRADABLE-INERT AND INSOLUBLE - MEETS LANDFILL REQUIREMENTS, (will not release liquids) -CERTIFIED FREE OF RESPIRABLE SILICA.
Catalog No. NC0100450 Supplier WYK Sorbents LLC Supplier No. S47443
Anti-Slip Safety Sorbent is concentrated and very economical to use. Studies have shown the average bag is used twelve times in cleaning up small spills and nuisance drips. A 510 bag will absorb up to 3 US gallons of oil and most other liquids.NON BIODEGRADABLE-INERT AND INSOLUBLE - MEETS LANDFILL REQUIREMENTS, (will not release liquids) -CERTIFIED FREE OF RESPIRABLE SILICA.