Learn More
Essendant Super Stitch Blend Mop Head, Medium, Cotton/Synthetic, Green, 6/Carton
Catalog No. 502004901
Encompass_Preferred
The Rubbermaid Commercial Super Stitch Blend Mop has a blend of post-industrial 4-ply cotton and synthetic fibers for higher absorbency and more launderings than cotton mops, and has looped ends and a yarn tailband to reduce tangling and fraying for improved performance. The synthetic mesh headband resists moisture retention so the mop head can dry between uses. Green cleaning equipment is recognized in the industry as color-coded for use in food service areas as part of a zone isolation system to help minimize cross-contamination. These mop heads are suitable for use in commercial and industrial applications.
Catalog No. 50-200-4901 Supplier Essendant Supplier No. RCPD252GRE
The Rubbermaid Commercial Super Stitch Blend Mop has a blend of post-industrial 4-ply cotton and synthetic fibers for higher absorbency and more launderings than cotton mops, and has looped ends and a yarn tailband to reduce tangling and fraying for improved performance. The synthetic mesh headband resists moisture retention so the mop head can dry between uses. Green cleaning equipment is recognized in the industry as color-coded for use in food service areas as part of a zone isolation system to help minimize cross-contamination. These mop heads are suitable for use in commercial and industrial applications.