Learn More
Sartorius Sartorius Super Streptavidin Biosensors, High Sensitivity, Regenerable, 96 biosensors
Supplier:  Sartorius 185057
Encompass
The Sartorius Octet Super Streptavidin Biosensors are designed for Biolayer Interferometry (BLI) applications, providing high sensitivity for small molecule kinetic analysis and fragment screening. These biosensors enable precise real-time measurements of kinetic interactions through reliable immobilization of biotinylated ligands. Their regenerable nature offers a cost-effective solution for high-throughput applications, differentiating them from standard biosensors.
Catalog No. NC9988411
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov