Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Learning Resources Super Strong Magnetic Hooks
SCI_ED

Catalog No. S02760
Change view
Click to view available options
Class Size: 1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S02760 Class Size: 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02760 Supplier Learning Resources Supplier No. LER2694
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Free up more wall and bulletin board space by storing materials on metal filing cabinets and other magnetic surfaces. Great for hanging pocket charts, flipbooks, decorations, hall passes, and more. Set of 4 comes in bright colors: red, blue, green and yellow.

  • Free up more wall and bulletin board space by storing materials on metal filing cabinets and other magnetic surfaces.
  • Great for hanging pocket charts, flipbooks, decorations, hall passes, and more.
  • Set of 4 comes in bright colors: red, blue, green and yellow.

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Product Type Magnetic Hooks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.