Learn More
Medchemexpress LLC Super-TDU (1-31) TFA | 99.3% | 3355.50 | 5 MG
SDP
Catalog No. 5000424217
Encompass_Preferred
Super-TDU (1-31) TFA is a peptide fragment derived from Super-TDU, functioning as an inhibitor of the YAP-TEAD complex. This compound demonstrates potent anti-tumor activity and effectively suppresses tumor growth in gastric cancer mouse models. It is intended for research use only and largely compromises increased cell viability induced by ACTN1 in liver cancer cell lines.
  • Inhibitor of YAP-TEAD complex.
  • Shows potent anti-tumor activity.
  • Suppresses tumor growth in gastric cancer mouse models.
  • Can compromise increased cell viability induced by ACTN1 in Huh-7 or LM3 cells proliferation.
  • Available in various quantities for research needs.
Catalog No. 50-004-24217 Supplier Medchemexpress LLC Supplier No. HYP1728A5MG
Super-TDU (1-31) TFA is a peptide fragment derived from Super-TDU, functioning as an inhibitor of the YAP-TEAD complex. This compound demonstrates potent anti-tumor activity and effectively suppresses tumor growth in gastric cancer mouse models. It is intended for research use only and largely compromises increased cell viability induced by ACTN1 in liver cancer cell lines.
  • Inhibitor of YAP-TEAD complex.
  • Shows potent anti-tumor activity.
  • Suppresses tumor growth in gastric cancer mouse models.
  • Can compromise increased cell viability induced by ACTN1 in Huh-7 or LM3 cells proliferation.
  • Available in various quantities for research needs.