Learn More
Medchemexpress LLC HY-P1727 10mg Medchemexpress, Super-TDU CAS:1599441-71-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP172710MG
Encompass_Preferred
Medchemexpress, HY-P1727 10mg Super-TDU CAS:1599441-71-0 Super-TDU is a specific YAP antagonist targeting YAP-TEADs interaction. Super-TDU suppresses tumor growth in gastric cancer mouse model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3072