Learn More
V&P Scientific Inc STIR ELEMENT, SUPER DISCS, Stainless Steel, Parylene C Coated, Used in Microplates, Syringes, Tubes/Vials, 1000/pack
Supplier:  V&P Scientific Inc VP 719F1
Encompass
STIR ELEMENT, SUPER DISCS, Stainless Steel, Parylene C Coated, 8.0mm Diameter, 0.7mm Height, 200°C Max Operating Temp for Coating
Catalog No. NC0576686