Learn More
MSC MetroVac Vacuum Cleaner, 8.0 Peak HP, 240 CFM, All Steel Construction
Catalog No. 502838329
Encompass
The SUPER VAC N BLASTER is a versatile unit designed for professional detailers and car enthusiasts, functioning as both a powerful vacuum and an efficient blower. It provides strong suction for thorough car interior cleaning and utilizes warm, dry, filtered air for safe, touchless drying, preventing scratches and streaks. Its durable all-steel construction ensures long-lasting performance.
  • Powerful suction generated by two 4.0 Peak HP twin-fan motors.
  • Blower function uses warm, dry, filtered air for touchless drying without scratching paint.
  • Clears water from inaccessible areas and blasts dust from AC vents and shift boots.
  • Can be used wall-mounted or as a portable unit with 10 ft. and 30 ft. commercial quality hoses.
  • Durable all-steel construction with a textured matte finish.
  • Includes a comprehensive set of accessories for detailing and organization.
Catalog No. 50-283-8329 Supplier MSC Supplier No. 59380444
The SUPER VAC N BLASTER is a versatile unit designed for professional detailers and car enthusiasts, functioning as both a powerful vacuum and an efficient blower. It provides strong suction for thorough car interior cleaning and utilizes warm, dry, filtered air for safe, touchless drying, preventing scratches and streaks. Its durable all-steel construction ensures long-lasting performance.
  • Powerful suction generated by two 4.0 Peak HP twin-fan motors.
  • Blower function uses warm, dry, filtered air for touchless drying without scratching paint.
  • Clears water from inaccessible areas and blasts dust from AC vents and shift boots.
  • Can be used wall-mounted or as a portable unit with 10 ft. and 30 ft. commercial quality hoses.
  • Durable all-steel construction with a textured matte finish.
  • Includes a comprehensive set of accessories for detailing and organization.