Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Super Vilella's Etchant 1% Picric Acid, 10% Hydrochloric Acid in Ethanol, Reagents

Super Vilella's Etchant 1% Picric Acid, 10% Hydrochloric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CV2003002.5D

Quantity 2.5 L

Catalog No. 01-333-398


 Restricted Item
Add to Cart

Description

Description

Specifications

Chemical Identifiers

64-17-5 , 7732-18-5 , 67-63-0
46.07
Vilellas
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol

Specifications

Yellow
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
Reagent
0.8276
2.5 L
Liquid
64-17-5 , 7732-18-5 , 67-63-0
Vilellas
CCO
46.07
Safety Coated Glass
0
Non-sterile
Super Vilella's Etchant 1% Picric Acid, 10% Hydrochloric Acid in Ethanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.