Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Super Yellow Light-emitting Ppv Copolymer

Catalog No. 900438500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-043-8500MG 500 mg
90-043-8250MG 250 mg
90-043-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 90-043-8500MG Supplier Sigma Aldrich Supplier No. 900438500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Super yellow light emitting PPV copolymer can form a thin film of poly(1,4-phenylenevinylene) (PPV) with high efficiencies. It has conjugated polymers as the backbone that can be soluble in water upon the incorporation of lipophilic solubilizing linkers.PDY-132 can be used in the preparation of electroluminescent line material, which can be used for making line art by direct-write patterning. It can also be used as a fluorescent material with the absorption peak at 450 nm, that can be developed by blade coating on the substrates of the transfer printed micro-sized light emitting diodes (++LED).Livilux is a trademark of Merck KGaA, Darmstadt, Germany

Specifications

CAS 26009-24-5
Quantity 500 mg
Synonym Livilux(TM); PDY-132
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.