Learn More
Bioworld Super Zymolyase, 2x0.5 mL, 1 mL
Catalog No. 50198997
Encompass_Preferred
Super Zymolyase A high performance Zymolyase preparation that is supplied in a liquid, ready to use form. The enzyme contains a high yeast lytic activity with low non-specific activity. Specifications:Suitable for yeast cell lysis, spheroplast preparation, glucan hyrolysis, etc. Stored at -20 C and used directly.2 x 0.5ml1.5u/ul
Catalog No. 50-198-997 Supplier Bioworld Supplier No. 215600291
Super Zymolyase A high performance Zymolyase preparation that is supplied in a liquid, ready to use form. The enzyme contains a high yeast lytic activity with low non-specific activity. Specifications:Suitable for yeast cell lysis, spheroplast preparation, glucan hyrolysis, etc. Stored at -20 C and used directly.2 x 0.5ml1.5u/ul