Learn More
MSC Loctite 234072 0.11 oz Tube Clear Instant Adhesive Series 495, 20 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Catalog No. 501113852
Encompass
Loctite 234072 0.11 oz Tube Clear Instant Adhesive Series 495, 20 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber
Catalog No. 50-111-3852 Supplier MSC Supplier No. 00256636
Loctite 234072 0.11 oz Tube Clear Instant Adhesive Series 495, 20 sec Fixture Time, 24 hr Full Cure Time, Bonds to Metal, Plastic & Rubber