Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperBoost™ Goat anti-Mouse Poly HRP

Catalog No. B40961
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40961 150 slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40961 Supplier Invitrogen™ Supplier No. B40961
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

SuperBoost™ Goat anti-Mouse Poly HRP is a component of SuperBoost™ tyramide signal amplification kits made available separately here.

SuperBoost™ Goat anti-Mouse Poly HRP can be used with any HRP substrate such as tyramide, DAB for signal amplification and detection. The molar enzyme/antibody protein ratio has an average value of '4'. This reagent is provided at a ready-to-use 1X concentration, enough for 150 slides.

When used as directed, this reagent along with other SuperBoost reagents can be used for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH) with up to 200 times more sensitivity than standard methods. For standout research, SuperBoost reagents sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal. SuperBoost reagents are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a SuperBoost reagent can be imaged using any type of microscope, producing high-resolution multiplex images.

Features of the SuperBoost reagents include:
• Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
• Simple protocol and detection using standard filters
• Suitable for high-resolution multiplex images—co-label with TO-PRO™-3, secondary antibodies, and other SuperBoost kits
• Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments

TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
Quantity 150 slides
Content And Storage 22.5 mL at 1X concentration in dropper bottle
Store at 2-8°C. When stored as directed, the product is stable for 6 months after receiving.
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Line SuperBoost

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.