Learn More
Protexer Corporation Shoe Cover Refill - SuperBootie - Case - 600 pairs
Catalog No. NC0854949
Encompass_Preferred
Completely waterproof and extra-durable. Constructed of spunbond polypropylene material with non-skid, non-conductive soles, with a polyethylene coating making it waterproof. Super anti-skid coating provides the ultimate in friction and traction. Strong elastic band around ankle allows for a universal fit. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.
This item is not returnable. View return policy
Catalog No. NC0854949 Supplier Protexer Corporation Supplier No. KBSB600
Completely waterproof and extra-durable. Constructed of spunbond polypropylene material with non-skid, non-conductive soles, with a polyethylene coating making it waterproof. Super anti-skid coating provides the ultimate in friction and traction. Strong elastic band around ankle allows for a universal fit. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.