Learn More
Protexer Corporation Shoe Cover Refill - SuperBootie with Rubber band - Case - 600 pairs
Catalog No. 501692601
Encompass_Preferred
Extra-durable. Constructed of spunbond polypropylene material with non-skid, nonconductive soles. Super anti-skid coating provides the ultimate in friction and traction. Strong elastic band around ankle allows for a universal fit. An added elastic band on bottom of shoe cover, allowing a more “Sure Fit” for users with smaller shoes/boot. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.
This item is not returnable. View return policy
Catalog No. 501692601 Supplier Protexer Corporation Supplier No. KBSBSF600
Extra-durable. Constructed of spunbond polypropylene material with non-skid, nonconductive soles. Super anti-skid coating provides the ultimate in friction and traction. Strong elastic band around ankle allows for a universal fit. An added elastic band on bottom of shoe cover, allowing a more “Sure Fit” for users with smaller shoes/boot. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.