Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SuperBroth Agar, Capsules

Catalog No. MP113010231 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity: 1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP113010231 Quantity: 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113010231 Supplier MP Biomedicals™ Supplier No. 113010231

missing translation for 'validMainframeMsg'

Culture media used for the growth of bacteria.

  • Used for bacteria cell culture, gene expression, transformation, microbiologics
TRUSTED_SUSTAINABILITY

Specifications

Format Capsules
Media Type Bacterial Media
Product Type Microbiology Media
Quantity 1 kg

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.