Learn More
Cayman Chemical Supercinnamaldehyde, Purity: ≥95%, CAS: 70351-51-8, 10 mg
SDP
Catalog No. 502984344
Encompass_Preferred
A TRPA1 channel activator (EC50 = 0.8 µM)
Catalog No. 50-298-4344 Supplier Cayman Chemical Supplier No. 2101910 MG
A TRPA1 channel activator (EC50 = 0.8 µM)